Assay Details
| Gene Symbol: cant1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003595.2 | NP_001003595.1 | 3 - 4 | 1028 | 59 | |
| XM_009306324.3 | XP_009304599.1 | 3 - 4 | 988 | 59 | ||
| XM_009306325.3 | XP_009304600.1 | 3 - 4 | 975 | 59 | ||
| GenBank mRNA | BC078222.1 | - | 3 - 4 | 1029 | 59 | |
More Information
Set Membership: |
|

cant1b
Genomic Map