Assay Details
Gene Symbol: syt9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003985.1 | NP_001003985.1 | 3 - 4 | 1269 | 85 | |
GenBank mRNA | BC080256.1 | - | 3 - 4 | 1269 | 85 | |
BC163971.1 | - | 3 - 4 | 1088 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
