Assay Details
| Gene Symbol: gpsm2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005936.1 | NP_001005936.1 | 11 - 12 | 1577 | 84 | |
| XM_021469788.1 | XP_021325463.1 | 11 - 12 | 1603 | 84 | ||
| GenBank mRNA | BC083520.1 | - | 11 - 12 | 1577 | 84 | |
| DQ235092.1 | - | 10 - 11 | 1424 | 84 | ||
More Information
Set Membership: |
|

gpsm2l
Genomic Map