Assay Details
| Gene Symbol: trpa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007066.2 | NP_001007067.1 | 27 - 28 | 3256 | 92 | |
| XM_009297331.3 | XP_009295606.1 | 27 - 28 | 3582 | 92 | ||
| GenBank mRNA | AY677197.1 | - | 27 - 28 | 3261 | 92 | |
| EU826642.1 | - | 26 - 27 | 3134 | 92 | ||
More Information
Set Membership: |
|

trpa1b
Genomic Map