Assay Details
| Gene Symbol: trpa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007066.2 | NP_001007067.1 | 9 - 10 | 1066 | 76 | |
| GenBank mRNA | AY677197.1 | - | 9 - 10 | 1071 | 76 | |
More Information
Set Membership: |
|

trpa1b
Genomic Map