Assay Details
Gene Symbol: gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007339.2 | NP_001007340.1 | 4 - 5 | 580 | 112 | |
GenBank mRNA | BC085540.1 | - | 4 - 5 | 583 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
