Assay Details
Gene Symbol: usp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008574.2 | NP_001008574.2 | 1 - 2 | 427 | 57 | |
XM_005157516.4 | XP_005157573.1 | 1 - 2 | 495 | 57 | ||
GenBank mRNA | BC086963.1 | - | 1 - 2 | 268 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
