Assay Details
Gene Symbol: usp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008574.2 | NP_001008574.2 | 3 - 4 | 604 | 73 | |
XM_005157513.4 | XP_005157570.1 | 5 - 6 | 1104 | 73 | ||
XM_005157514.4 | XP_005157571.1 | 5 - 6 | 1198 | 73 | ||
XM_005157515.4 | XP_005157572.1 | 5 - 6 | 1099 | 73 | ||
XM_005157516.4 | XP_005157573.1 | 3 - 4 | 672 | 73 | ||
XM_017350951.2 | XP_017206440.1 | 5 - 6 | 1099 | 73 | ||
XM_021481396.1 | XP_021337071.1 | 5 - 6 | 1202 | 73 | ||
GenBank mRNA | BC086963.1 | - | 3 - 4 | 445 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
