Assay Details
Gene Symbol: usp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008574.2 | NP_001008574.2 | 6 - 7 | 888 | 115 | |
XM_005157513.4 | XP_005157570.1 | 8 - 9 | 1388 | 115 | ||
XM_005157514.4 | XP_005157571.1 | 8 - 9 | 1482 | 115 | ||
XM_005157515.4 | XP_005157572.1 | 8 - 9 | 1383 | 115 | ||
XM_005157516.4 | XP_005157573.1 | 6 - 7 | 956 | 115 | ||
XM_017350951.2 | XP_017206440.1 | 8 - 9 | 1383 | 115 | ||
XM_021481396.1 | XP_021337071.1 | 8 - 9 | 1486 | 115 | ||
GenBank mRNA | BC086963.1 | - | 6 - 7 | 729 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
