Assay Details
Gene Symbol: usp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008574.2 | NP_001008574.2 | 7 - 8 | 993 | 84 | |
XM_005157515.4 | XP_005157572.1 | 9 - 10 | 1488 | 84 | ||
GenBank mRNA | BC086963.1 | - | 7 - 8 | 834 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
