Assay Details
| Gene Symbol: fgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009561.1 | NP_001009561.1 | 1 - 2 | 220 | 55 | |
| GenBank mRNA | AB194699.1 | - | 1 - 2 | 220 | 55 | |
| BC162921.1 | - | 1 - 2 | 242 | 55 | ||
| BC162923.1 | - | 1 - 2 | 242 | 55 | ||
More Information
Set Membership: |
|

fgf5
Genomic Map