Assay Details
Gene Symbol: crygs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012262.1 | NP_001012262.1 | 2 - 3 | 293 | 78 | |
GenBank mRNA | AY738744.1 | - | 2 - 3 | 293 | 78 | |
BC162365.1 | - | 2 - 3 | 293 | 78 | ||
BC162374.1 | - | 2 - 3 | 293 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
