Assay Details
| Gene Symbol: apobec2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013314.2 | NP_001013332.1 | 1 - 2 | 240 | 117 | |
| GenBank mRNA | BC090758.1 | - | 1 - 2 | 244 | 117 | |
| BC165110.1 | - | 1 - 2 | 264 | 117 | ||
| FJ469677.1 | - | 1 - 2 | 230 | 117 | ||
More Information
Set Membership: |
|

apobec2a
Genomic Map