Assay Details
| Gene Symbol: apobec2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013314.2 | NP_001013332.1 | 2 - 3 | 765 | 74 | |
| GenBank mRNA | BC090758.1 | - | 2 - 3 | 769 | 74 | |
More Information
Set Membership: |
|

apobec2a
Genomic Map