Assay Details
Gene Symbol: c6ast3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013526.1 | NP_001013544.1 | 9 - 10 | 829 | 60 | |
GenBank mRNA | BC090904.1 | - | 9 - 10 | 829 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
