Assay Details
| Gene Symbol: rbm39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014370.1 | NP_001014392.1 | 10 - 11 | 1175 | 102 | |
| XM_005167222.4 | XP_005167279.1 | 9 - 10 | 1125 | 102 | ||
| XM_005167224.4 | XP_005167281.1 | 10 - 11 | 1200 | 102 | ||
| XM_009304133.3 | XP_009302408.1 | 11 - 12 | 1346 | 102 | ||
| XM_021478249.1 | XP_021333924.1 | 9 - 10 | 1138 | 102 | ||
| XM_021478250.1 | XP_021333925.1 | 9 - 10 | 1026 | 102 | ||
| XM_021478251.1 | XP_021333926.1 | 9 - 10 | 1125 | 102 | ||
| GenBank mRNA | BC091794.1 | - | 10 - 11 | 1175 | 102 | |
More Information
Set Membership: |
|

rbm39b
Genomic Map