Assay Details
| Gene Symbol: rbm39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014370.1 | NP_001014392.1 | 12 - 13 | 1437 | 64 | |
| XM_005167222.4 | XP_005167279.1 | 11 - 12 | 1387 | 64 | ||
| XM_005167224.4 | XP_005167281.1 | 12 - 13 | 1462 | 64 | ||
| XM_009304133.3 | XP_009302408.1 | 13 - 14 | 1608 | 64 | ||
| XM_021478249.1 | XP_021333924.1 | 11 - 12 | 1400 | 64 | ||
| XM_021478250.1 | XP_021333925.1 | 11 - 12 | 1288 | 64 | ||
| GenBank mRNA | BC091794.1 | - | 12 - 13 | 1437 | 64 | |
More Information
Set Membership: |
|

rbm39b
Genomic Map