Assay Details
| Gene Symbol: srd5a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017693.1 | NP_001017693.1 | 3 - 4 | 589 | 77 | |
| XM_005156994.4 | XP_005157051.1 | 3 - 4 | 671 | 77 | ||
| GenBank mRNA | BC093319.1 | - | 3 - 4 | 589 | 77 | |
| BC164314.1 | - | 3 - 4 | 535 | 77 | ||
More Information
Set Membership: |
|

srd5a2b
Genomic Map