Assay Details
| Gene Symbol: syt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020546.2 | NP_001018382.2 | 2 - 3 | 282 | 81 | |
| GenBank mRNA | BC095109.1 | - | 2 - 3 | 282 | 81 | |
| BC165563.1 | - | 1 - 2 | 199 | 81 | ||
More Information
Set Membership: |
|

syt5b
Genomic Map