Assay Details
| Gene Symbol: mrps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020559.2 | NP_001018395.1 | 2 - 3 | 294 | 84 | |
| GenBank mRNA | BC095143.1 | - | 2 - 3 | 297 | 84 | |
More Information
Set Membership: |
|

mrps17
Genomic Map