Assay Details
Gene Symbol: acbd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020647.1 | NP_001018483.1 | 3 - 4 | 274 | 55 | |
XM_005162509.4 | XP_005162566.1 | 3 - 4 | 296 | 55 | ||
XM_009297238.3 | XP_009295513.1 | 3 - 4 | 296 | 55 | ||
XM_009297239.3 | XP_009295514.1 | 3 - 4 | 281 | 55 | ||
XM_009297241.3 | XP_009295516.1 | 3 - 4 | 338 | 55 | ||
XM_021470173.1 | XP_021325848.1 | 3 - 4 | 438 | 55 | ||
XM_021470174.1 | XP_021325849.1 | 3 - 4 | 296 | 55 | ||
GenBank mRNA | BC095655.1 | - | 3 - 4 | 274 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
