Assay Details
Gene Symbol: pak2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025456.1 | NP_001020627.1 | 4 - 5 | 749 | 72 | |
XM_005170360.4 | XP_005170417.1 | 4 - 5 | 626 | 72 | ||
GenBank mRNA | BC081414.1 | - | 4 - 5 | 606 | 72 | |
BC095627.1 | - | 4 - 5 | 720 | 72 | ||
BC096967.1 | - | 4 - 5 | 749 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
