Assay Details
| Gene Symbol: ppp6r2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025509.1 | NP_001020680.1 | 10 - 11 | 1702 | 90 | |
| XM_021475043.1 | XP_021330718.1 | 9 - 10 | 1495 | 90 | ||
| GenBank mRNA | BC115199.1 | - | 10 - 11 | 1586 | 90 | |
More Information
Set Membership: |
|

ppp6r2a
Genomic Map