Assay Details
Gene Symbol: rtn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029965.3 | NP_001025136.1 | 3 - 4 | 432 | 76 | |
XM_005157471.4 | XP_005157528.1 | 6 - 7 | 1241 | 76 | ||
XM_005157472.4 | XP_005157529.1 | 3 - 4 | 480 | 76 | ||
GenBank mRNA | AY576807.1 | - | 3 - 4 | 432 | 76 | |
BC115054.1 | - | 3 - 4 | 430 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
