Assay Details
Gene Symbol: rtn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029965.3 | NP_001025136.1 | 4 - 5 | 501 | 67 | |
XM_005157471.4 | XP_005157528.1 | 7 - 8 | 1310 | 67 | ||
XM_005157472.4 | XP_005157529.1 | 4 - 5 | 549 | 67 | ||
GenBank mRNA | AY576807.1 | - | 4 - 5 | 501 | 67 | |
BC115054.1 | - | 4 - 5 | 499 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
