Assay Details
| Gene Symbol: bmp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039812.1 | NP_001034901.1 | 18 - 19 | 2714 | 98 | |
| XM_005155406.4 | XP_005155463.1 | 18 - 19 | 2768 | 98 | ||
| GenBank mRNA | AB239772.1 | - | 18 - 19 | 2714 | 98 | |
More Information
Set Membership: |
|

bmp1b
Genomic Map