Assay Details
| Gene Symbol: retsatl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040004.2 | NP_001035093.2 | 7 - 8 | 1359 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

retsatl| Gene Symbol: retsatl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040004.2 | NP_001035093.2 | 7 - 8 | 1359 | 72 | |
Set Membership: |
|