Assay Details
| Gene Symbol: bmp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040037.1 | NP_001035126.1 | 8 - 9 | 1076 | 106 | |
| XM_017357365.2 | XP_017212854.1 | 8 - 9 | 1098 | 106 | ||
| GenBank mRNA | AB239771.1 | - | 8 - 9 | 1076 | 106 | |
| BC163535.1 | - | 8 - 9 | 1064 | 106 | ||
More Information
Set Membership: |
|

bmp1a
Genomic Map