Assay Details
Gene Symbol: cacna2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044960.2 | NP_001038425.2 | 19 - 20 | 1763 | 87 | |
XM_005164746.4 | XP_005164803.1 | 19 - 20 | 1939 | 87 | ||
XM_009300285.3 | XP_009298560.1 | 19 - 20 | 1941 | 87 | ||
XM_009300286.3 | XP_009298561.1 | 19 - 20 | 1941 | 87 | ||
XM_009300287.3 | XP_009298562.1 | 19 - 20 | 1939 | 87 | ||
XM_009300288.3 | XP_009298563.1 | 19 - 20 | 1941 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
