Assay Details
Gene Symbol: cacna2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044960.2 | NP_001038425.2 | 28 - 29 | 2466 | 89 | |
XM_005164746.4 | XP_005164803.1 | 28 - 29 | 2642 | 89 | ||
XM_009300285.3 | XP_009298560.1 | 29 - 30 | 2668 | 89 | ||
XM_009300286.3 | XP_009298561.1 | 29 - 30 | 2665 | 89 | ||
XM_009300287.3 | XP_009298562.1 | 29 - 30 | 2666 | 89 | ||
XM_009300288.3 | XP_009298563.1 | 28 - 29 | 2644 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
