Assay Details
| Gene Symbol: crb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045162.1 | NP_001038627.1 | 10 - 11 | 3251 | 71 | |
| XM_017357360.2 | XP_017212849.1 | 10 - 11 | 3293 | 71 | ||
| GenBank mRNA | DQ314737.1 | - | 10 - 11 | 3251 | 71 | |
| JQ740885.1 | - | 4 - 5 | 1883 | 71 | ||
More Information
Set Membership: |
|

crb2b
Genomic Map