Assay Details
| Gene Symbol: crb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045162.1 | NP_001038627.1 | 13 - 14 | 4374 | 113 | |
| XM_017357360.2 | XP_017212849.1 | 13 - 14 | 4416 | 113 | ||
| GenBank mRNA | DQ314737.1 | - | 13 - 14 | 4374 | 113 | |
| JQ740885.1 | - | 7 - 8 | 3006 | 113 | ||
More Information
Set Membership: |
|

crb2b
Genomic Map