Assay Details
| Gene Symbol: crb2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045299.1 | NP_001038764.1 | 8 - 9 | 3083 | 61 | |
| GenBank mRNA | BC162451.1 | - | 8 - 9 | 3065 | 61 | |
| CU638819.1 | - | 8 - 9 | 2903 | 61 | ||
| DQ314736.1 | - | 8 - 9 | 3083 | 61 | ||
More Information
Set Membership: |
|

crb2a
Genomic Map