Assay Details
| Gene Symbol: rab43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045347.1 | NP_001038812.1 | 2 - 3 | 508 | 68 | |
| XM_005155678.4 | XP_005155735.1 | 3 - 4 | 653 | 68 | ||
| XM_005155679.4 | XP_005155736.1 | 3 - 4 | 659 | 68 | ||
| GenBank mRNA | BC122421.1 | - | 2 - 3 | 508 | 68 | |
| BC165016.1 | - | 2 - 3 | 401 | 68 | ||
More Information
Set Membership: |
|

rab43
Genomic Map