Assay Details
| Gene Symbol: kti12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126418.1 | NP_001119890.1 | 3 - 4 | 366 | 119 | |
| GenBank mRNA | BC122265.1 | - | 3 - 4 | 356 | 119 | |
| BC164624.1 | - | 3 - 4 | 291 | 119 | ||
More Information
Set Membership: |
|

kti12
Genomic Map