Assay Details
| Gene Symbol: kti12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126418.1 | NP_001119890.1 | 8 - 9 | 793 | 76 | |
| GenBank mRNA | BC122265.1 | - | 8 - 9 | 783 | 76 | |
| BC164624.1 | - | 8 - 9 | 718 | 76 | ||
More Information
Set Membership: |
|

kti12
Genomic Map