Assay Details
Gene Symbol: csnk1g2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045850.2 | NP_001039315.1 | 1 - 2 | 781 | 87 | |
NM_001290336.1 | NP_001277265.1 | 2 - 3 | 912 | 87 | ||
XM_005166859.4 | XP_005166916.1 | 2 - 3 | 946 | 87 | ||
GenBank mRNA | BC122280.1 | - | 1 - 2 | 349 | 87 | |
BC164532.1 | - | 1 - 2 | 256 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
