Assay Details
| Gene Symbol: csnk1g2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045850.2 | NP_001039315.1 | 9 - 10 | 1725 | 150 | |
| XM_005166859.4 | XP_005166916.1 | 10 - 11 | 1890 | 150 | ||
| GenBank mRNA | BC122280.1 | - | 9 - 10 | 1293 | 150 | |
| BC164532.1 | - | 9 - 10 | 1200 | 150 | ||
More Information
Set Membership: |
|

csnk1g2b
Genomic Map