Assay Details
Gene Symbol: gtf2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076571.1 | NP_001070039.1 | 5 - 6 | 458 | 64 | |
XM_017358801.2 | XP_017214290.1 | 5 - 6 | 723 | 64 | ||
GenBank mRNA | BC124541.1 | - | 5 - 6 | 458 | 64 | |
BC164474.1 | - | 5 - 6 | 421 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
