Assay Details
Gene Symbol: med19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076591.1 | NP_001070059.1 | 2 - 3 | 431 | 66 | |
GenBank mRNA | BC124344.1 | - | 2 - 3 | 431 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
