Assay Details
Gene Symbol: pafah1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082851.1 | NP_001076320.1 | 3 - 4 | 281 | 88 | |
XM_005165344.4 | XP_005165401.1 | 4 - 5 | 636 | 88 | ||
XM_021476285.1 | XP_021331960.1 | 5 - 6 | 381 | 88 | ||
GenBank mRNA | BC160649.1 | - | 4 - 5 | 607 | 88 | |
BC163819.1 | - | 3 - 4 | 305 | 88 | ||
BC163859.1 | - | 3 - 4 | 305 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
