Assay Details
Gene Symbol: pcsk5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083829.1 | NP_001077298.1 | 8 - 9 | 1395 | 82 | |
GenBank mRNA | BC162145.1 | - | 8 - 9 | 1253 | 82 | |
DQ983233.1 | - | 8 - 9 | 1395 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
