Assay Details
Gene Symbol: rock2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093747.2 | NP_001087216.1 | 14 - 15 | 1654 | 66 | |
XM_009294695.3 | XP_009292970.1 | 14 - 15 | 2126 | 66 | ||
XM_017352433.2 | XP_017207922.1 | 14 - 15 | 2127 | 66 | ||
XM_017352434.2 | XP_017207923.1 | 14 - 15 | 2127 | 66 | ||
XM_017352437.2 | XP_017207926.1 | 14 - 15 | 2130 | 66 | ||
XM_021468356.1 | XP_021324031.1 | 14 - 15 | 2129 | 66 | ||
XM_021468357.1 | XP_021324032.1 | 14 - 15 | 2128 | 66 | ||
XM_021468358.1 | XP_021324033.1 | 14 - 15 | 2128 | 66 | ||
XM_021468359.1 | XP_021324034.1 | 14 - 15 | 2128 | 66 | ||
XM_021468360.1 | XP_021324035.1 | 14 - 15 | 2131 | 66 | ||
XM_021468361.1 | XP_021324036.1 | 14 - 15 | 2129 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
