Assay Details
Gene Symbol: kiss1rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110531.1 | NP_001104001.1 | 4 - 5 | 742 | 130 | |
XM_005168656.4 | XP_005168713.1 | 5 - 6 | 1341 | 130 | ||
GenBank mRNA | EU047918.1 | - | 4 - 5 | 742 | 130 | |
HQ222884.1 | - | 3 - 4 | 617 | 130 | ||
HQ222887.1 | - | 4 - 5 | 825 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
