Assay Details
| Gene Symbol: ccl27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328381.1 | NP_001315310.1 | 2 - 3 | 220 | 80 | |
| NM_131062.2 | NP_571137.2 | 2 - 3 | 217 | 80 | ||
| GenBank mRNA | AF201450.1 | - | 2 - 3 | 204 | 80 | |
| EH445572.1 | - | 2 - 3 | 230 | 80 | ||
| EH456016.1 | - | 2 - 3 | 240 | 80 | ||
More Information
Set Membership: |
|

ccl27a
Genomic Map