Assay Details
Gene Symbol: rxrba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131275.1 | NP_571350.1 | 11 - 12 | 1364 | 76 | |
XM_005159410.3 | XP_005159467.1 | 10 - 11 | 1363 | 76 | ||
XM_005159411.3 | XP_005159468.1 | 9 - 10 | 1339 | 76 | ||
XM_009294022.2 | XP_009292297.1 | 11 - 12 | 1390 | 76 | ||
XM_009294023.2 | XP_009292298.1 | 10 - 11 | 1366 | 76 | ||
XM_009294026.2 | XP_009292301.1 | 11 - 12 | 1412 | 76 | ||
XM_021467983.1 | XP_021323658.1 | 12 - 13 | 1439 | 76 | ||
GenBank mRNA | BC054649.1 | - | 10 - 11 | 1189 | 76 | |
U29942.1 | - | 11 - 12 | 1364 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
