Assay Details
| Gene Symbol: rxrba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131275.1 | NP_571350.1 | 13 - 14 | 1553 | 70 | |
| XM_005159410.3 | XP_005159467.1 | 12 - 13 | 1552 | 70 | ||
| XM_005159411.3 | XP_005159468.1 | 11 - 12 | 1528 | 70 | ||
| XM_005159412.3 | XP_005159469.1 | 10 - 11 | 1486 | 70 | ||
| XM_009294022.2 | XP_009292297.1 | 13 - 14 | 1579 | 70 | ||
| XM_009294023.2 | XP_009292298.1 | 12 - 13 | 1555 | 70 | ||
| XM_009294024.2 | XP_009292299.1 | 12 - 13 | 1537 | 70 | ||
| XM_009294025.2 | XP_009292300.1 | 11 - 12 | 1513 | 70 | ||
| XM_009294026.2 | XP_009292301.1 | 13 - 14 | 1601 | 70 | ||
| XM_021467983.1 | XP_021323658.1 | 14 - 15 | 1628 | 70 | ||
| GenBank mRNA | U29942.1 | - | 13 - 14 | 1553 | 70 | |
More Information
Set Membership: |
|

rxrba
Genomic Map