Assay Details
| Gene Symbol: rxrba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131275.1 | NP_571350.1 | 4 - 5 | 537 | 64 | |
| XM_005159410.3 | XP_005159467.1 | 3 - 4 | 536 | 64 | ||
| XM_005159411.3 | XP_005159468.1 | 3 - 4 | 536 | 64 | ||
| XM_005159412.3 | XP_005159469.1 | 3 - 4 | 536 | 64 | ||
| XM_009294026.2 | XP_009292301.1 | 4 - 5 | 585 | 64 | ||
| GenBank mRNA | BC054649.1 | - | 3 - 4 | 362 | 64 | |
| EF519314.1 | - | 4 - 5 | 453 | 64 | ||
| EF519315.1 | - | 3 - 4 | 404 | 64 | ||
| U29942.1 | - | 4 - 5 | 537 | 64 | ||
More Information
Set Membership: |
|

rxrba
Genomic Map