Assay Details
Gene Symbol: rxrba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131275.1 | NP_571350.1 | 7 - 8 | 899 | 108 | |
XM_005159410.3 | XP_005159467.1 | 6 - 7 | 898 | 108 | ||
XM_009294022.2 | XP_009292297.1 | 7 - 8 | 925 | 108 | ||
XM_009294024.2 | XP_009292299.1 | 7 - 8 | 925 | 108 | ||
XM_009294026.2 | XP_009292301.1 | 7 - 8 | 947 | 108 | ||
XM_021467983.1 | XP_021323658.1 | 8 - 9 | 974 | 108 | ||
GenBank mRNA | U29942.1 | - | 7 - 8 | 899 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
