Assay Details
| Gene Symbol: rxrba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131275.1 | NP_571350.1 | 9 - 10 | 1185 | 107 | |
| XM_005159410.3 | XP_005159467.1 | 8 - 9 | 1184 | 107 | ||
| XM_005159411.3 | XP_005159468.1 | 7 - 8 | 1160 | 107 | ||
| XM_005159412.3 | XP_005159469.1 | 7 - 8 | 1160 | 107 | ||
| XM_009294022.2 | XP_009292297.1 | 9 - 10 | 1211 | 107 | ||
| XM_009294023.2 | XP_009292298.1 | 8 - 9 | 1187 | 107 | ||
| XM_009294024.2 | XP_009292299.1 | 9 - 10 | 1211 | 107 | ||
| XM_009294025.2 | XP_009292300.1 | 8 - 9 | 1187 | 107 | ||
| XM_009294026.2 | XP_009292301.1 | 9 - 10 | 1233 | 107 | ||
| XM_021467983.1 | XP_021323658.1 | 10 - 11 | 1260 | 107 | ||
| GenBank mRNA | BC054649.1 | - | 8 - 9 | 1010 | 107 | |
| U29942.1 | - | 9 - 10 | 1185 | 107 | ||
More Information
Set Membership: |
|

rxrba
Genomic Map